Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605143 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACTR1A - ARP1 actin-related protein 1 homolog A, centractin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFSD13A - major facilitator superfamily domain containing 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024789.3 | Intron | NP_079065.2 | ||||
XM_006717973.3 | Intron | XP_006718036.1 | ||||
XM_006717974.3 | Intron | XP_006718037.1 | ||||
XM_011540160.2 | Intron | XP_011538462.1 | ||||
XM_011540163.2 | Intron | XP_011538465.1 | ||||
XM_011540167.2 | Intron | XP_011538469.1 | ||||
XM_011540169.2 | Intron | XP_011538471.1 | ||||
XM_017016654.1 | Intron | XP_016872143.1 | ||||
XM_017016655.1 | Intron | XP_016872144.1 | ||||
XM_017016656.1 | Intron | XP_016872145.1 | ||||
XM_017016657.1 | Intron | XP_016872146.1 | ||||
XM_017016658.1 | Intron | XP_016872147.1 | ||||
XM_017016659.1 | Intron | XP_016872148.1 | ||||
XM_017016660.1 | Intron | XP_016872149.1 | ||||
XM_017016661.1 | Intron | XP_016872150.1 | ||||
XM_017016662.1 | Intron | XP_016872151.1 |