Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607680 | ||||||||||||||||||||
Literature Links: |
RCHY1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RCHY1 - ring finger and CHY zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009922.2 | Intron | NP_001009922.1 | ||||
NM_001278536.1 | Intron | NP_001265465.1 | ||||
NM_001278537.1 | Intron | NP_001265466.1 | ||||
NM_001278538.1 | Intron | NP_001265467.1 | ||||
NM_001278539.1 | Intron | NP_001265468.1 | ||||
NM_015436.3 | Intron | NP_056251.2 | ||||
XM_011531838.2 | Intron | XP_011530140.1 | ||||
XM_011531839.2 | Intron | XP_011530141.1 |