Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600789 MIM: 600692 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL23 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MRPL23 - mitochondrial ribosomal protein L23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021134.3 | Intron | NP_066957.3 | ||||
XM_006718271.2 | Intron | XP_006718334.1 | ||||
XM_011520273.1 | Intron | XP_011518575.1 | ||||
XM_011520275.2 | Intron | XP_011518577.1 | ||||
XM_011520276.2 | Intron | XP_011518578.1 |
SNORD131 - small nucleolar RNA, C/D box 131 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNT3 - troponin T3, fast skeletal type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |