Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602992 | ||||||||||||||||||||
Literature Links: |
LAIR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAIR1 - leukocyte associated immunoglobulin like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289023.2 | 1632 | UTR 3 | NP_001275952.2 | |||
NM_001289025.2 | 1632 | UTR 3 | NP_001275954.2 | |||
NM_001289026.2 | 1632 | UTR 3 | NP_001275955.2 | |||
NM_001289027.2 | 1632 | UTR 3 | NP_001275956.2 | |||
NM_002287.5 | 1632 | UTR 3 | NP_002278.2 | |||
NM_021706.4 | 1632 | UTR 3 | NP_068352.2 | |||
XM_017026803.1 | 1632 | Intron | XP_016882292.1 |