Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606398 MIM: 609742 MIM: 605815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF5 - activating transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193646.1 | 467 | Silent Mutation | CTC,CTT | L,L 30 | NP_001180575.1 | |
NM_001290746.1 | 467 | Silent Mutation | CTC,CTT | L,L 30 | NP_001277675.1 | |
NM_012068.5 | 467 | Silent Mutation | CTC,CTT | L,L 30 | NP_036200.2 | |
XM_011526629.2 | 467 | Silent Mutation | CTC,CTT | L,L 30 | XP_011524931.1 |
IL4I1 - interleukin 4 induced 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4751 - microRNA 4751 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP62 - nucleoporin 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |