Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612465 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBC1D4 - TBC1 domain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286658.2 | 6353 | UTR 3 | NP_001273587.1 | |||
NM_001286659.2 | 6353 | UTR 3 | NP_001273588.1 | |||
NM_014832.4 | 6353 | UTR 3 | NP_055647.2 | |||
XM_005266603.1 | 6353 | UTR 3 | XP_005266660.1 | |||
XM_005266605.2 | 6353 | UTR 3 | XP_005266662.1 | |||
XM_006719903.3 | 6353 | UTR 3 | XP_006719966.1 | |||
XM_011535331.1 | 6353 | UTR 3 | XP_011533633.1 | |||
XM_017020882.1 | 6353 | UTR 3 | XP_016876371.1 | |||
XM_017020883.1 | 6353 | UTR 3 | XP_016876372.1 | |||
XM_017020884.1 | 6353 | UTR 3 | XP_016876373.1 |