Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613904 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF569 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF569 - zinc finger protein 569 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF570 - zinc finger protein 570 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300993.2 | Intron | NP_001287922.1 | ||||
NM_001321991.1 | Intron | NP_001308920.1 | ||||
NM_001321992.1 | Intron | NP_001308921.1 | ||||
NM_001321993.1 | Intron | NP_001308922.1 | ||||
NM_001321994.1 | Intron | NP_001308923.1 | ||||
NM_144694.3 | Intron | NP_653295.1 | ||||
XM_011526544.2 | Intron | XP_011524846.1 | ||||
XM_011526545.2 | Intron | XP_011524847.1 |
Set Membership: |
HapMap |