Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCAGTTTTACTGGCCCATCCT[A/T]GAGGAGCTTAATGTCACTCGGCCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607611 MIM: 608076 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLSCR3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PLSCR3 - phospholipid scramblase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM256-PLSCR3 - TMEM256-PLSCR3 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNK1 - tyrosine kinase non receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251902.1 | 313 | Silent Mutation | CTA,CTT | L,L 27 | NP_001238831.1 | |
NM_003985.4 | 313 | Silent Mutation | CTA,CTT | L,L 27 | NP_003976.2 | |
XM_011524045.2 | 313 | Silent Mutation | CTA,CTT | L,L 27 | XP_011522347.1 | |
XM_011524046.2 | 313 | Silent Mutation | CTA,CTT | L,L 27 | XP_011522348.1 | |
XM_011524047.2 | 313 | Silent Mutation | CTA,CTT | L,L 27 | XP_011522349.1 | |
XM_011524048.1 | 313 | UTR 5 | XP_011522350.1 |