Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615496 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
||||||
SAS - Not Available | Japanese
|
JPT (Japanese)
|
||||||
AFR - Not Available | Chinese
|
CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
AGBL1 - ATP/GTP binding protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011521226.2 | Intron | XP_011519528.1 | ||||
XM_011521227.2 | Intron | XP_011519529.1 | ||||
XM_011521228.2 | Intron | XP_011519530.1 | ||||
XM_011521230.2 | Intron | XP_011519532.1 | ||||
XM_017021918.1 | Intron | XP_016877407.1 | ||||
XM_017021919.1 | Intron | XP_016877408.1 | ||||
XM_017021920.1 | Intron | XP_016877409.1 | ||||
XM_017021921.1 | Intron | XP_016877410.1 |
LINC01584 - long intergenic non-protein coding RNA 1584 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |