Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147370 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF1R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IGF1R - insulin like growth factor 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000875.4 | Intron | NP_000866.1 | ||||
NM_001291858.1 | Intron | NP_001278787.1 | ||||
XM_011521516.2 | Intron | XP_011519818.1 | ||||
XM_011521517.2 | Intron | XP_011519819.1 | ||||
XM_017022136.1 | Intron | XP_016877625.1 | ||||
XM_017022137.1 | Intron | XP_016877626.1 | ||||
XM_017022138.1 | Intron | XP_016877627.1 | ||||
XM_017022139.1 | Intron | XP_016877628.1 |
IRAIN - IGF1R antisense imprinted non-protein coding RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |