Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137960 MIM: 606474 MIM: 600945 | ||||||||||||||||||||
Literature Links: |
MPV17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPV17 - MPV17, mitochondrial inner membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002437.4 | 935 | UTR 3 | NP_002428.1 | |||
XM_005264326.3 | 935 | UTR 3 | XP_005264383.1 | |||
XM_006712021.3 | 935 | UTR 3 | XP_006712084.1 | |||
XM_017004150.1 | 935 | UTR 3 | XP_016859639.1 | |||
XM_017004151.1 | 935 | UTR 3 | XP_016859640.1 | |||
XM_017004152.1 | 935 | UTR 3 | XP_016859641.1 |
TRIM54 - tripartite motif containing 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCN - urocortin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |