Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606010 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NRBP1 - nuclear receptor binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321357.1 | Intron | NP_001308286.1 | ||||
NM_001321358.1 | Intron | NP_001308287.1 | ||||
NM_001321359.1 | Intron | NP_001308288.1 | ||||
NM_001321361.1 | Intron | NP_001308290.1 | ||||
NM_001321362.1 | Intron | NP_001308291.1 | ||||
NM_001321363.1 | Intron | NP_001308292.1 | ||||
NM_013392.3 | Intron | NP_037524.1 |
Set Membership: |
HapMap |