Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603182 MIM: 606106 | ||||||||||||||||||||
Literature Links: |
ILF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ILF3 - interleukin enhancer binding factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137673.1 | 120 | UTR 5 | NP_001131145.1 | |||
NM_004516.3 | 120 | UTR 5 | NP_004507.2 | |||
NM_012218.3 | 120 | UTR 5 | NP_036350.2 | |||
NM_017620.2 | 120 | UTR 5 | NP_060090.2 | |||
NM_153464.2 | 120 | UTR 5 | NP_703194.1 | |||
XM_006722742.2 | 120 | UTR 5 | XP_006722805.1 | |||
XM_011527984.2 | 120 | Intron | XP_011526286.2 | |||
XM_011527985.2 | 120 | UTR 5 | XP_011526287.1 | |||
XM_011527987.1 | 120 | Intron | XP_011526289.1 | |||
XM_017026763.1 | 120 | UTR 5 | XP_016882252.1 | |||
XM_017026764.1 | 120 | Intron | XP_016882253.1 | |||
XM_017026765.1 | 120 | UTR 5 | XP_016882254.1 |
ILF3-AS1 - ILF3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC44A2 - solute carrier family 44 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |