Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPC102 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSPC102 - uncharacterized HSPC102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN16 - tetraspanin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282509.1 | Intron | NP_001269438.1 | ||||
NM_001282510.1 | Intron | NP_001269439.1 | ||||
NM_012466.3 | Intron | NP_036598.1 | ||||
XM_011527901.1 | Intron | XP_011526203.1 | ||||
XM_011527902.1 | Intron | XP_011526204.1 | ||||
XM_011527903.2 | Intron | XP_011526205.1 |
Set Membership: |
HapMap |