Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCACTAGAAGGTGGAGAAGGAG[A/G]GCGGGAAGTCCAGTTGAGATGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600806 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNN1 - calponin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELOF1 - elongation factor 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032377.3 | 963 | UTR 3 | NP_115753.1 | |||
XM_017027356.1 | 963 | UTR 3 | XP_016882845.1 | |||
XM_017027357.1 | 963 | UTR 3 | XP_016882846.1 | |||
XM_017027358.1 | 963 | UTR 3 | XP_016882847.1 | |||
XM_017027359.1 | 963 | UTR 3 | XP_016882848.1 |
LOC105369211 - uncharacterized LOC105369211 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |