Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139320 MIM: 610540 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNAS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GNAS - GNAS complex locus | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000516.5 | 375 | Intron | NP_000507.1 | |||
NM_001077488.3 | 375 | Intron | NP_001070956.1 | |||
NM_001077489.3 | 375 | Intron | NP_001070957.1 | |||
NM_001077490.2 | 375 | Intron | NP_001070958.1 | |||
NM_001309840.1 | 375 | Intron | NP_001296769.1 | |||
NM_001309842.1 | 375 | Intron | NP_001296771.1 | |||
NM_001309861.1 | 375 | Intron | NP_001296790.1 | |||
NM_001309883.1 | 375 | Intron | NP_001296812.1 | |||
NM_016592.3 | 375 | UTR 5 | NP_057676.1 | |||
NM_080425.3 | 375 | Intron | NP_536350.2 | |||
NM_080426.3 | 375 | Intron | NP_536351.1 | |||
XM_017027812.1 | 375 | Intron | XP_016883301.1 | |||
XM_017027813.1 | 375 | Intron | XP_016883302.1 | |||
XM_017027814.1 | 375 | Intron | XP_016883303.1 | |||
XM_017027815.1 | 375 | Intron | XP_016883304.1 | |||
XM_017027816.1 | 375 | Intron | XP_016883305.1 | |||
XM_017027817.1 | 375 | Intron | XP_016883306.1 | |||
XM_017027818.1 | 375 | Intron | XP_016883307.1 | |||
XM_017027819.1 | 375 | Intron | XP_016883308.1 | |||
XM_017027820.1 | 375 | Intron | XP_016883309.1 | |||
XM_017027821.1 | 375 | UTR 5 | XP_016883310.1 | |||
XM_017027822.1 | 375 | UTR 5 | XP_016883311.1 |
GNAS-AS1 - GNAS antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |