Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607758 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTNNBIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CTNNBIP1 - catenin beta interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012329.1 | 2048 | UTR 3 | NP_001012329.1 | |||
NM_020248.2 | 2048 | UTR 3 | NP_064633.1 | |||
XM_017001853.1 | 2048 | UTR 3 | XP_016857342.1 | |||
XM_017001854.1 | 2048 | UTR 3 | XP_016857343.1 | |||
XM_017001855.1 | 2048 | UTR 3 | XP_016857344.1 |