Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609385 MIM: 142810 | ||||||||||||||||||||
Literature Links: |
DND1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DND1 - DND microRNA-mediated repression inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194249.2 | 4019 | Intron | NP_919225.1 |
HARS - histidyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258040.2 | 4019 | UTR 3 | NP_001244969.1 | |||
NM_001258041.2 | 4019 | UTR 3 | NP_001244970.1 | |||
NM_001258042.2 | 4019 | UTR 3 | NP_001244971.1 | |||
NM_001289092.1 | 4019 | UTR 3 | NP_001276021.1 | |||
NM_001289093.1 | 4019 | UTR 3 | NP_001276022.1 | |||
NM_001289094.1 | 4019 | UTR 3 | NP_001276023.1 | |||
NM_002109.5 | 4019 | UTR 3 | NP_002100.2 |
WDR55 - WD repeat domain 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017706.4 | 4019 | Intron | NP_060176.2 | |||
XM_005268469.2 | 4019 | UTR 3 | XP_005268526.1 | |||
XM_017009600.1 | 4019 | UTR 3 | XP_016865089.1 |