Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP78 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP78 - centrosomal protein 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098802.1 | Intron | NP_001092272.1 | ||||
NM_032171.1 | Intron | NP_115547.1 | ||||
XM_005252263.4 | Intron | XP_005252320.1 | ||||
XM_005252265.2 | Intron | XP_005252322.1 | ||||
XM_005252266.4 | Intron | XP_005252323.1 | ||||
XM_017015192.1 | Intron | XP_016870681.1 | ||||
XM_017015193.1 | Intron | XP_016870682.1 | ||||
XM_017015194.1 | Intron | XP_016870683.1 | ||||
XM_017015195.1 | Intron | XP_016870684.1 | ||||
XM_017015196.1 | Intron | XP_016870685.1 |