Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602688 MIM: 614683 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPAB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HNRNPAB - heterogeneous nuclear ribonucleoprotein A/B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004499.3 | 1514 | UTR 3 | NP_004490.2 | |||
NM_031266.2 | 1514 | UTR 3 | NP_112556.2 |
PHYKPL - 5-phosphohydroxy-L-lysine phospho-lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278346.1 | 1514 | Intron | NP_001265275.1 | |||
NM_153373.3 | 1514 | Intron | NP_699204.1 | |||
XM_006714925.2 | 1514 | Intron | XP_006714988.1 | |||
XM_011534669.2 | 1514 | Intron | XP_011532971.1 | |||
XM_011534670.2 | 1514 | Intron | XP_011532972.1 | |||
XM_011534671.2 | 1514 | Intron | XP_011532973.1 | |||
XM_011534672.2 | 1514 | Intron | XP_011532974.1 | |||
XM_011534674.2 | 1514 | Intron | XP_011532976.1 | |||
XM_011534676.2 | 1514 | Intron | XP_011532978.1 | |||
XM_017009995.1 | 1514 | Intron | XP_016865484.1 | |||
XM_017009996.1 | 1514 | Intron | XP_016865485.1 | |||
XM_017009997.1 | 1514 | Intron | XP_016865486.1 | |||
XM_017009998.1 | 1514 | Intron | XP_016865487.1 | |||
XM_017009999.1 | 1514 | Intron | XP_016865488.1 |