Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGACACCCAGGCCTCCTGGCGT[A/C]AGTGGGGCCTGGGACGTCTGGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC100507336 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507336 - uncharacterized LOC100507336 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PXDC1 - PX domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183373.3 | 1304 | UTR 3 | NP_899229.2 | |||
XM_011514393.2 | 1304 | UTR 3 | XP_011512695.1 |