Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610661 | ||||||||||||||||||||
Literature Links: |
NGLY1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NGLY1 - N-glycanase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145293.1 | 1966 | UTR 3 | NP_001138765.1 | |||
NM_001145294.1 | 1966 | UTR 3 | NP_001138766.1 | |||
NM_001145295.1 | 1966 | UTR 3 | NP_001138767.1 | |||
NM_018297.3 | 1966 | UTR 3 | NP_060767.2 | |||
XM_005265316.1 | 1966 | UTR 3 | XP_005265373.1 | |||
XM_005265317.1 | 1966 | UTR 3 | XP_005265374.1 | |||
XM_011533944.1 | 1966 | UTR 3 | XP_011532246.1 | |||
XM_011533945.2 | 1966 | Intron | XP_011532247.1 | |||
XM_017006839.1 | 1966 | Intron | XP_016862328.1 |