Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606573 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRK PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRK - fyn related Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002031.2 | 2301 | UTR 3 | NP_002022.1 | |||
XM_005266880.4 | 2301 | UTR 3 | XP_005266937.1 | |||
XM_005266881.2 | 2301 | UTR 3 | XP_005266938.1 | |||
XM_005266882.4 | 2301 | UTR 3 | XP_005266939.1 | |||
XM_011535653.2 | 2301 | UTR 3 | XP_011533955.1 | |||
XM_011535654.2 | 2301 | UTR 3 | XP_011533956.1 | |||
XM_011535655.2 | 2301 | UTR 3 | XP_011533957.1 | |||
XM_011535656.2 | 2301 | UTR 3 | XP_011533958.1 | |||
XM_017010645.1 | 2301 | UTR 3 | XP_016866134.1 |