Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUSD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SUSD1 - sushi domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282640.1 | 2420 | UTR 3 | NP_001269569.1 | |||
NM_001282643.1 | 2420 | UTR 3 | NP_001269572.1 | |||
NM_022486.4 | 2420 | Silent Mutation | NP_071931.2 | |||
XM_011518915.1 | 2420 | UTR 3 | XP_011517217.1 | |||
XM_011518916.2 | 2420 | Silent Mutation | XP_011517218.1 | |||
XM_011518917.1 | 2420 | UTR 3 | XP_011517219.1 | |||
XM_011518918.1 | 2420 | UTR 3 | XP_011517220.1 | |||
XM_011518919.1 | 2420 | UTR 3 | XP_011517221.1 | |||
XM_017015037.1 | 2420 | Silent Mutation | XP_016870526.1 | |||
XM_017015038.1 | 2420 | UTR 3 | XP_016870527.1 | |||
XM_017015039.1 | 2420 | UTR 3 | XP_016870528.1 | |||
XM_017015040.1 | 2420 | Intron | XP_016870529.1 |
Set Membership: |
HapMap |