Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610558 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPKAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAPKAP1 - mitogen-activated protein kinase associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006617.1 | 3087 | UTR 3 | NP_001006618.1 | |||
NM_001006618.1 | 3087 | Intron | NP_001006619.1 | |||
NM_001006619.1 | 3087 | UTR 3 | NP_001006620.1 | |||
NM_001006620.1 | 3087 | UTR 3 | NP_001006621.1 | |||
NM_001006621.1 | 3087 | UTR 3 | NP_001006622.1 | |||
NM_024117.3 | 3087 | UTR 3 | NP_077022.1 | |||
XM_011519005.1 | 3087 | UTR 3 | XP_011517307.1 | |||
XM_011519006.2 | 3087 | Intron | XP_011517308.1 | |||
XM_011519007.1 | 3087 | UTR 3 | XP_011517309.1 | |||
XM_011519008.1 | 3087 | Intron | XP_011517310.1 | |||
XM_011519009.1 | 3087 | Intron | XP_011517311.1 | |||
XM_011519010.2 | 3087 | Intron | XP_011517312.1 | |||
XM_017015126.1 | 3087 | UTR 3 | XP_016870615.1 | |||
XM_017015127.1 | 3087 | Intron | XP_016870616.1 |
Set Membership: |
HapMap Validated |