Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608215 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHX6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LHX6 - LIM homeobox 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242333.1 | Intron | NP_001229262.1 | ||||
NM_001242334.1 | Intron | NP_001229263.1 | ||||
NM_001242335.1 | Intron | NP_001229264.1 | ||||
NM_014368.4 | Intron | NP_055183.2 | ||||
NM_199160.3 | Intron | NP_954629.2 | ||||
XM_005251916.3 | Intron | XP_005251973.1 | ||||
XM_011518521.2 | Intron | XP_011516823.1 | ||||
XM_011518522.2 | Intron | XP_011516824.1 |
MORN5 - MORN repeat containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286828.1 | Intron | NP_001273757.1 | ||||
NM_198469.3 | Intron | NP_940871.2 | ||||
XM_005251877.4 | Intron | XP_005251934.1 | ||||
XM_005251878.3 | Intron | XP_005251935.1 | ||||
XM_006717034.3 | Intron | XP_006717097.1 | ||||
XM_011518457.2 | Intron | XP_011516759.1 | ||||
XM_011518458.1 | Intron | XP_011516760.1 | ||||
XM_017014578.1 | Intron | XP_016870067.1 |
Set Membership: |
HapMap Validated |