Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605712 | ||||||||||||||||||||
Literature Links: |
SPTLC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPTLC1 - serine palmitoyltransferase long chain base subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281303.1 | 1604 | Silent Mutation | CTG,TTG | L,L 490 | NP_001268232.1 | |
NM_006415.3 | 1604 | UTR 3 | NP_006406.1 | |||
NM_178324.2 | 1604 | Intron | NP_847894.1 | |||
XM_011518138.2 | 1604 | Intron | XP_011516440.1 | |||
XM_011518139.2 | 1604 | UTR 3 | XP_011516441.1 | |||
XM_017014200.1 | 1604 | UTR 3 | XP_016869689.1 | |||
XM_017014201.1 | 1604 | UTR 3 | XP_016869690.1 | |||
XM_017014202.1 | 1604 | UTR 3 | XP_016869691.1 |