Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142704 MIM: 603247 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HDC - histidine decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306146.1 | 1825 | Silent Mutation | AAC,AAT | N,N 505 | NP_001293075.1 | |
NM_002112.3 | 1825 | Silent Mutation | AAC,AAT | N,N 538 | NP_002103.2 | |
XM_017022094.1 | 1825 | Silent Mutation | AAC,AAT | N,N 573 | XP_016877583.1 | |
XM_017022095.1 | 1825 | Silent Mutation | AAC,AAT | N,N 540 | XP_016877584.1 | |
XM_017022096.1 | 1825 | Silent Mutation | AAC,AAT | N,N 497 | XP_016877585.1 | |
XM_017022097.1 | 1825 | Silent Mutation | AAC,AAT | N,N 494 | XP_016877586.1 | |
XM_017022098.1 | 1825 | Silent Mutation | AAC,AAT | N,N 429 | XP_016877587.1 | |
XM_017022099.1 | 1825 | Intron | XP_016877588.1 |
SLC27A2 - solute carrier family 27 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |