Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGGGGAGGACTGTGGGGCTGT[A/G]GATTCGTGGCGTTTTCTGTTCTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603958 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287944 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100287944 - uncharacterized LOC100287944 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFX4 - regulatory factor X4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206691.1 | Intron | NP_001193620.1 | ||||
NM_032491.5 | Intron | NP_115880.2 | ||||
NM_213594.2 | Intron | NP_998759.1 |
Set Membership: |
HapMap |