Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603201 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ABCB11 - ATP binding cassette subfamily B member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003742.2 | 3184 | UTR 3 | NP_003733.2 | |||
XM_006712817.3 | 3184 | UTR 3 | XP_006712880.1 | |||
XM_011512077.2 | 3184 | UTR 3 | XP_011510379.1 | |||
XM_011512078.2 | 3184 | UTR 3 | XP_011510380.1 | |||
XM_011512080.2 | 3184 | Intron | XP_011510382.1 | |||
XM_011512081.2 | 3184 | UTR 3 | XP_011510383.1 | |||
XM_017005165.1 | 3184 | Intron | XP_016860654.1 | |||
XM_017005166.1 | 3184 | UTR 3 | XP_016860655.1 | |||
XM_017005167.1 | 3184 | UTR 3 | XP_016860656.1 |
Set Membership: |
HapMap DME Validated Inventoried |