Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600040 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAX PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BAX - BCL2 associated X, apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291428.1 | Intron | NP_001278357.1 | ||||
NM_001291429.1 | Intron | NP_001278358.1 | ||||
NM_001291430.1 | Intron | NP_001278359.1 | ||||
NM_001291431.1 | Intron | NP_001278360.1 | ||||
NM_004324.3 | Intron | NP_004315.1 | ||||
NM_138761.3 | Intron | NP_620116.1 | ||||
NM_138763.3 | Intron | NP_620118.1 | ||||
NM_138764.4 | Intron | NP_620119.2 | ||||
XM_017027077.1 | Intron | XP_016882566.1 |