Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATGGGCCTTGGACAATCACTCA[C/T]GTACCCAAATGTGTTCTCCAGATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613948 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPATA8 - spermatogenesis associated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304804.1 | Intron | NP_001291733.1 | ||||
NM_001304805.1 | Intron | NP_001291734.1 | ||||
NM_001304806.1 | Intron | NP_001291735.1 | ||||
NM_173499.4 | Intron | NP_775770.1 | ||||
XM_011521265.2 | Intron | XP_011519567.1 |
SPATA8-AS1 - SPATA8 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |