Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602651 MIM: 606737 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRDC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NRDC - nardilysin convertase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101662.1 | Intron | NP_001095132.1 | ||||
NM_001242361.1 | Intron | NP_001229290.1 | ||||
NM_002525.2 | Intron | NP_002516.2 | ||||
XM_005270903.1 | Intron | XP_005270960.1 | ||||
XM_011541522.1 | Intron | XP_011539824.1 | ||||
XM_011541523.1 | Intron | XP_011539825.1 | ||||
XM_011541525.2 | Intron | XP_011539827.1 | ||||
XM_017001375.1 | Intron | XP_016856864.1 |
Set Membership: |
HapMap |