Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601540 | ||||||||||||||||||||
Literature Links: |
BRD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRD2 - bromodomain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113182.2 | 647 | UTR 5 | NP_001106653.1 | |||
NM_001199455.1 | 647 | UTR 5 | NP_001186384.1 | |||
NM_001199456.1 | 647 | Intron | NP_001186385.1 | |||
NM_001291986.1 | 647 | UTR 5 | NP_001278915.1 | |||
NM_005104.3 | 647 | UTR 5 | NP_005095.1 |