Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120290 MIM: 601417 MIM: 180246 MIM: 601416 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL11A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL11A2 - collagen type XI alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSD17B8 - hydroxysteroid 17-beta dehydrogenase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RXRB - retinoid X receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270401.1 | 1322 | Silent Mutation | TTC,TTT | F,F 384 | NP_001257330.1 | |
NM_001291989.1 | 1322 | Silent Mutation | TTC,TTT | F,F 194 | NP_001278918.1 | |
NM_021976.4 | 1322 | Silent Mutation | TTC,TTT | F,F 384 | NP_068811.1 | |
XM_005249278.2 | 1322 | Silent Mutation | TTC,TTT | F,F 288 | XP_005249335.1 | |
XM_011514796.2 | 1322 | Silent Mutation | TTC,TTT | F,F 267 | XP_011513098.1 | |
XM_017011176.1 | 1322 | Silent Mutation | TTC,TTT | F,F 261 | XP_016866665.1 |
SLC39A7 - solute carrier family 39 member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |