Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604716 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGT2A1 - UDP glucuronosyltransferase family 2 member A1 complex locus | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252274.2 | Intron | NP_001239203.2 | ||||
NM_001252275.2 | Intron | NP_001239204.2 | ||||
NM_001301239.1 | Intron | NP_001288168.1 | ||||
NM_006798.4 | Intron | NP_006789.3 |
UGT2A2 - UDP glucuronosyltransferase family 2 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105677.2 | Intron | NP_001099147.2 | ||||
NM_001301233.1 | Intron | NP_001288162.1 |