Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
70 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603944 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1614 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA1614 - KIAA1614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX6 - syntaxin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286210.1 | Intron | NP_001273139.1 | ||||
NM_005819.5 | Intron | NP_005810.1 | ||||
XM_011509058.1 | Intron | XP_011507360.1 | ||||
XM_011509059.1 | Intron | XP_011507361.1 | ||||
XM_017000006.1 | Intron | XP_016855495.1 | ||||
XM_017000007.1 | Intron | XP_016855496.1 |
Set Membership: |
HapMap |