Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604388 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GNG4 - G protein subunit gamma 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098721.1 | 637 | UTR 3 | NP_001092191.1 | |||
NM_001098722.1 | 637 | UTR 3 | NP_001092192.1 | |||
NM_004485.3 | 637 | UTR 3 | NP_004476.1 | |||
XM_006711761.2 | 637 | UTR 3 | XP_006711824.1 | |||
XM_011544167.2 | 637 | UTR 3 | XP_011542469.1 |
Set Membership: |
HapMap JSNP |