Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
76 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605102 MIM: 605078 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MASP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MASP2 - mannan binding lectin serine peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006610.3 | 2318 | UTR 3 | NP_006601.2 | |||
NM_139208.2 | 2318 | Intron | NP_631947.1 | |||
XM_017000097.1 | 2318 | Intron | XP_016855586.1 |
TARDBP - TAR DNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007375.3 | 2318 | Intron | NP_031401.1 | |||
XM_017000863.1 | 2318 | Intron | XP_016856352.1 | |||
XM_017000864.1 | 2318 | Intron | XP_016856353.1 | |||
XM_017000865.1 | 2318 | Intron | XP_016856354.1 | |||
XM_017000866.1 | 2318 | Intron | XP_016856355.1 | |||
XM_017000867.1 | 2318 | Intron | XP_016856356.1 | |||
XM_017000868.1 | 2318 | Intron | XP_016856357.1 |
Set Membership: |
HapMap |