Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612640 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBATA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TBATA - thymus, brain and testes associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318241.1 | Intron | NP_001305170.1 | ||||
NM_001318242.1 | Intron | NP_001305171.1 | ||||
NM_001318243.1 | Intron | NP_001305172.1 | ||||
NM_152710.3 | Intron | NP_689923.3 | ||||
XM_017015845.1 | Intron | XP_016871334.1 | ||||
XM_017015846.1 | Intron | XP_016871335.1 | ||||
XM_017015847.1 | Intron | XP_016871336.1 | ||||
XM_017015848.1 | Intron | XP_016871337.1 | ||||
XM_017015849.1 | Intron | XP_016871338.1 | ||||
XM_017015850.1 | Intron | XP_016871339.1 | ||||
XM_017015851.1 | Intron | XP_016871340.1 | ||||
XM_017015852.1 | Intron | XP_016871341.1 | ||||
XM_017015853.1 | Intron | XP_016871342.1 | ||||
XM_017015854.1 | Intron | XP_016871343.1 | ||||
XM_017015855.1 | Intron | XP_016871344.1 | ||||
XM_017015856.1 | Intron | XP_016871345.1 | ||||
XM_017015857.1 | Intron | XP_016871346.1 | ||||
XM_017015858.1 | Intron | XP_016871347.1 | ||||
XM_017015859.1 | Intron | XP_016871348.1 | ||||
XM_017015860.1 | Intron | XP_016871349.1 | ||||
XM_017015861.1 | Intron | XP_016871350.1 | ||||
XM_017015862.1 | Intron | XP_016871351.1 |