Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACTCAGTTTCACCTTGGATAAA[A/G]AAAGCCATTTGTTAAATGTAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNJ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNJ - cyclin J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134375.1 | Intron | NP_001127847.1 | ||||
NM_001134376.1 | Intron | NP_001127848.1 | ||||
NM_019084.4 | Intron | NP_061957.2 | ||||
XM_005269921.3 | Intron | XP_005269978.1 | ||||
XM_011539885.2 | Intron | XP_011538187.1 | ||||
XM_017016354.1 | Intron | XP_016871843.1 | ||||
XM_017016355.1 | Intron | XP_016871844.1 | ||||
XM_017016356.1 | Intron | XP_016871845.1 |
ENTPD1-AS1 - ENTPD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |