Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608412 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GPBP1 - GC-rich promoter binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127235.2 | Intron | NP_001120707.2 | ||||
NM_001127236.2 | Intron | NP_001120708.1 | ||||
NM_001203246.1 | Intron | NP_001190175.1 | ||||
NM_022913.3 | Intron | NP_075064.1 | ||||
XM_006714673.3 | Intron | XP_006714736.1 | ||||
XM_006714674.3 | Intron | XP_006714737.1 | ||||
XM_017009754.1 | Intron | XP_016865243.1 | ||||
XM_017009755.1 | Intron | XP_016865244.1 | ||||
XM_017009756.1 | Intron | XP_016865245.1 | ||||
XM_017009757.1 | Intron | XP_016865246.1 | ||||
XM_017009758.1 | Intron | XP_016865247.1 | ||||
XM_017009759.1 | Intron | XP_016865248.1 |