Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGAAGTCTGGAGTCTCCAGG[C/T]GGGCCTGGCACGTCAGCGTCACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138670 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
A1BG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
A1BG - alpha-1-B glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130786.3 | 217 | Missense Mutation | CAC,CGC | H,R 52 | NP_570602.2 |
A1BG-AS1 - A1BG antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100419840 - zinc finger protein 446 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372483 - uncharacterized LOC105372483 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF497 - zinc finger protein 497 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |