Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613183 MIM: 609281 | ||||||||||||||||||||
Literature Links: |
BOLA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BOLA3 - bolA family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BOLA3-AS1 - BOLA3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOB1A - MOB kinase activator 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317110.1 | 2026 | UTR 3 | NP_001304039.1 | |||
NM_001317111.1 | 2026 | UTR 3 | NP_001304040.1 | |||
NM_001317112.1 | 2026 | UTR 3 | NP_001304041.1 | |||
NM_018221.4 | 2026 | UTR 3 | NP_060691.2 | |||
XM_011532948.2 | 2026 | UTR 3 | XP_011531250.1 |