Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGTGAGAGGCCAGAACCGCAG[C/T]GGGGAGTAAAGGTGAGCTGGGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610559 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RASSF4 - Ras association domain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032023.3 | Intron | NP_114412.2 | ||||
XM_005271833.4 | Intron | XP_005271890.1 | ||||
XM_005271835.4 | Intron | XP_005271892.1 | ||||
XM_006718013.3 | Intron | XP_006718076.1 | ||||
XM_006718014.3 | Intron | XP_006718077.1 | ||||
XM_011540255.2 | Intron | XP_011538557.1 | ||||
XM_017016754.1 | Intron | XP_016872243.1 | ||||
XM_017016755.1 | Intron | XP_016872244.1 | ||||
XM_017016756.1 | Intron | XP_016872245.1 | ||||
XM_017016757.1 | Intron | XP_016872246.1 | ||||
XM_017016758.1 | Intron | XP_016872247.1 |
TMEM72-AS1 - TMEM72 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |