Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608277 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHST15 - carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270764.1 | 3696 | UTR 3 | NP_001257693.1 | |||
NM_001270765.1 | 3696 | Intron | NP_001257694.1 | |||
NM_014863.3 | 3696 | Intron | NP_055678.2 | |||
NM_015892.4 | 3696 | UTR 3 | NP_056976.2 | |||
XM_005269891.3 | 3696 | UTR 3 | XP_005269948.1 | |||
XM_005269892.4 | 3696 | Intron | XP_005269949.1 | |||
XM_005269893.3 | 3696 | Intron | XP_005269950.1 | |||
XM_005269894.4 | 3696 | Intron | XP_005269951.1 | |||
XM_006717891.3 | 3696 | UTR 3 | XP_006717954.1 | |||
XM_011539857.1 | 3696 | Intron | XP_011538159.1 | |||
XM_017016319.1 | 3696 | UTR 3 | XP_016871808.1 | |||
XM_017016320.1 | 3696 | Intron | XP_016871809.1 | |||
XM_017016321.1 | 3696 | Intron | XP_016871810.1 |
Set Membership: |
HapMap |