Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613349 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OAT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OAT - ornithine aminotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000274.3 | Intron | NP_000265.1 | ||||
NM_001171814.1 | Intron | NP_001165285.1 | ||||
NM_001322965.1 | Intron | NP_001309894.1 | ||||
NM_001322966.1 | Intron | NP_001309895.1 | ||||
NM_001322967.1 | Intron | NP_001309896.1 | ||||
NM_001322968.1 | Intron | NP_001309897.1 | ||||
NM_001322969.1 | Intron | NP_001309898.1 | ||||
NM_001322970.1 | Intron | NP_001309899.1 | ||||
NM_001322971.1 | Intron | NP_001309900.1 | ||||
NM_001322974.1 | Intron | NP_001309903.1 | ||||
XM_017016279.1 | Intron | XP_016871768.1 |
Set Membership: |
HapMap Validated |