Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120110 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL10A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL10A1 - collagen type X alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000493.3 | 6612 | UTR 3 | NP_000484.2 | |||
XM_006715333.3 | 6612 | UTR 3 | XP_006715396.1 | |||
XM_011535432.2 | 6612 | UTR 3 | XP_011533734.1 | |||
XM_011535433.2 | 6612 | UTR 3 | XP_011533735.1 | |||
XM_017010248.1 | 6612 | UTR 3 | XP_016865737.1 | |||
XM_017010249.1 | 6612 | UTR 3 | XP_016865738.1 |
NT5DC1 - 5'-nucleotidase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152729.2 | 6612 | Intron | NP_689942.2 | |||
XM_006715378.3 | 6612 | Intron | XP_006715441.1 |
Set Membership: |
HapMap |