Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSANTD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MSANTD4 - Myb/SANT DNA binding domain containing 4 with coiled-coils | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318747.1 | 2897 | UTR 3 | NP_001305676.1 | |||
NM_001318748.1 | 2897 | UTR 3 | NP_001305677.1 | |||
NM_001318749.1 | 2897 | UTR 3 | NP_001305678.1 | |||
NM_001318750.1 | 2897 | UTR 3 | NP_001305679.1 | |||
NM_032424.2 | 2897 | UTR 3 | NP_115800.1 | |||
XM_011543022.2 | 2897 | UTR 3 | XP_011541324.1 | |||
XM_017018416.1 | 2897 | UTR 3 | XP_016873905.1 | |||
XM_017018417.1 | 2897 | UTR 3 | XP_016873906.1 |