Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615855 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OVCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OVCH1 - ovochymase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMTC1 - transmembrane and tetratricopeptide repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193451.1 | 8567 | UTR 3 | NP_001180380.1 | |||
NM_175861.3 | 8567 | UTR 3 | NP_787057.2 | |||
XM_005253499.4 | 8567 | UTR 3 | XP_005253556.1 | |||
XM_017020003.1 | 8567 | UTR 3 | XP_016875492.1 | |||
XM_017020004.1 | 8567 | UTR 3 | XP_016875493.1 | |||
XM_017020005.1 | 8567 | UTR 3 | XP_016875494.1 | |||
XM_017020006.1 | 8567 | Intron | XP_016875495.1 | |||
XM_017020007.1 | 8567 | UTR 3 | XP_016875496.1 |